Please login to your online account to display your discounted pricing

anti-Contactin 6 Polyclonal, Thermo Scientific™

Contactin 6 Polyclonal antibody PA1-18250 from Thermo Scientific™ specifically detects Contactin 6 in Rat samples, and it is validated for Immunohistochemistry

$543.75 - $543.75

Specifications

Antigen Contactin 6
Cross Reactivity Rat
Format Serum
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues G Y P S P H Y R W K Q of mouse contactin-6.
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

01-670-503

 
pierce biotechnology inc
PA118250
Unlabeled 100μL Each for $543.75

01-670-504

 
pierce biotechnology inc
PA118251
Unlabeled 100μL Each for $543.75
Description & Specifications

Specifications

Antigen Contactin 6
Cross Reactivity Rat
Format Serum
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues G Y P S P H Y R W K Q of mouse contactin-6.
Isotype IgG
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Storage Requirements Avoid Freeze/Thaw Cycles

Contactin 6 Polyclonal antibody PA1-18250 from Thermo Scientific™ specifically detects Contactin 6 in Rat samples, and it is validated for Immunohistochemistry

The neural adhesion molecule Contactin-6, also known as NB-3, is a contactin/F3 subgroup member of immunoglobulin superfamily. It is expressed exclusively in the nervous system and mainly upregulated at the early postnatal stage during mouse brain development.
Host Species: Rabbit
Cross Reactivity: Rat
Isotype: IgG
Immunogen: Synthetic peptide corresponding to residues G Y P S P H Y R W K Q of mouse contactin-6.
Formulation: 100μL unpurified Antibody in whole serum, with no preservative;
Synonyms: Neural recognition molecule NB-3, CNTN6
Uniprot Accession: P97528
Gene Symbol: Cntn6
Entrez GeneID: 27256
PA1-18250 detects Contactin 6 in Rat samples. PA1-18250 has been successfully used in Immunohistochemistry procedures. PA1-18250 The immunogen is Synthetic peptide corresponding to residues G Y P S P H Y R W K Q of mouse contactin-6..