Please login to your online account to display your discounted pricing

anti-Contactin 5 Polyclonal, Thermo Scientific™


Contactin 5 Polyclonal antibody OSC00015W from Thermo Scientific™ specifically detects Contactin 5 in Human samples, and it is validated for Immunohistochemistry and Western Blot

Manufacturer: pierce biotechnology inc  OSC00015W

 View more versions of this product

Catalog No. PIOSC00015W

  • / Each
 

Description & Specifications

Specifications

Formulation 100μL unpurified antibody in whole serum and no preservative
Applications Immunohistochemistry
Applications Western Blot
Gene ID (Entrez) 53942
Cross Reactivity Human
Format Serum
Species Reactivity Human
Gene Accession No. O94779
Gene Alias Contactin 5, Neural recognition molecule NB-2, NB-2, hNB-2, HNB-2s, CNTN5, MGC163491
Gene Symbols CNTN5
Storage Requirements avoiding freeze/thaw cycles.
Host Species Rabbit
Immunogen A synthetic peptide from human contactin-5
Quantity 100μL
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled
Antigen Contactin 5

Contactin 5 Polyclonal antibody OSC00015W from Thermo Scientific™ specifically detects Contactin 5 in Human samples, and it is validated for Immunohistochemistry and Western Blot


Host Species: Rabbit
Cross Reactivity: Human
Immunogen: A synthetic peptide from human contactin-5
Formulation: 100μL unpurified antibody in whole serum and no preservative
Synonyms: Contactin 5, Neural recognition molecule NB-2, NB-2, hNB-2, HNB-2s, CNTN5, MGC163491
Uniprot Accession: O94779
Entrez GeneID: 53942
Gene Symbol: CNTN5
Reconstitute with 100 ul of distilled water.