Please login to your online account to display your discounted pricing

anti-Contactin 5 Polyclonal, Thermo Scientific™

Contactin 5 Polyclonal antibody OSC00015W from Thermo Scientific™ specifically detects Contactin 5 in Human samples, and it is validated for Immunohistochemistry and Western Blot

$448.05 - $448.05

Specifications

Antigen Contactin 5
Cross Reactivity Rat
Format Serum
Host Species Rabbit
Immunogen A synthetic peptide from rat Contactin-5
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

PIOSC00019W

 
pierce biotechnology inc
OSC00019W
Unlabeled 100μL Each for $448.05

PIOSC00021W

 
pierce biotechnology inc
OSC00021W
Unlabeled 100μL Each for $448.05

PIOSC00015W

 
pierce biotechnology inc
OSC00015W
Unlabeled 100μL Each for $448.05
Description & Specifications

Specifications

Antigen Contactin 5
Cross Reactivity Rat
Format Serum
Host Species Rabbit
Immunogen A synthetic peptide from rat Contactin-5
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Storage Requirements avoiding freeze/thaw cycles.

Contactin 5 Polyclonal antibody OSC00015W from Thermo Scientific™ specifically detects Contactin 5 in Human samples, and it is validated for Immunohistochemistry and Western Blot


Host Species: Rabbit
Cross Reactivity: Human
Immunogen: A synthetic peptide from human contactin-5
Formulation: 100μL unpurified Antibody in whole serum, with no preservative;
Synonyms: Contactin 5, Neural recognition molecule NB-2, NB-2, hNB-2, HNB-2s, CNTN5, MGC163491
Uniprot Accession: O94779
Gene Symbol: CNTN5
Entrez GeneID: 53942
OSC00015W detects Contactin 5 in Human samples. OSC00015W has been successfully used in Immunohistochemistry and Western Blot procedures. OSC00015W The immunogen is A synthetic peptide from human contactin-5.