Please login to your online account to display your discounted pricing

anti-Contactin 5 Polyclonal, Thermo Scientific™


Contactin 5 Polyclonal antibody OSC00015W from Thermo Scientific™ specifically detects Contactin 5 in Human samples, and it is validated for Immunohistochemistry and Western Blot

Manufacturer: pierce biotechnology inc  OSC00021W

 View more versions of this product

Catalog No. PIOSC00021W

  • / Each
 

Description & Specifications

Specifications

Formulation 100μL unpurified antibody in whole serum and no preservative
Gene Symbols CNTN5
Applications Immunohistochemistry
Applications Western Blot
Gene ID (Entrez) 114589
Cross Reactivity Rat
Gene Accession No. P97527
Storage Requirements avoiding freeze/thaw cycles.
Format Serum
Gene Alias NB-2, contactin 5, CNTN5, Neural recognition molecule NB-2
Species Reactivity Rat
Quantity 100μL
Antigen Contactin 5
Host Species Rabbit
Immunogen A synthetic peptide from rat Contactin-5
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled

Contactin 5 Polyclonal antibody OSC00015W from Thermo Scientific™ specifically detects Contactin 5 in Human samples, and it is validated for Immunohistochemistry and Western Blot


Host Species: Rabbit
Cross Reactivity: Rat
Immunogen: A synthetic peptide from rat Contactin-5
Formulation: 100μL unpurified antibody in whole serum and no preservative
Synonyms: NB-2, contactin 5, CNTN5, Neural recognition molecule NB-2
Uniprot Accession: P97527
Entrez GeneID: 114589
Gene Symbol: CNTN5
Reconstitute with 100 ul of distilled water.