Please login to your online account to display your discounted pricing

Contactin 5 Polyclonal Antibody, Invitrogen™


Contactin 5 Antibody, OSC00019W, from Invitrogen™. Species Reactivity: Mouse; Applications: Immunohistochemistry, Western Blot.

Manufacturer: pierce biotechnology inc  OSC00019W

Catalog No. PIOSC00019W

  • / Each
 

Description & Specifications

Specifications

Formulation 100μL unpurified antibody in whole serum and no preservative
Gene Symbols CNTN5
Applications Immunohistochemistry
Applications Western Blot
Gene ID (Entrez) 244682
Cross Reactivity Mouse
Gene Accession No. P68500
Storage Requirements avoiding freeze/thaw cycles.
Format Serum
Gene Alias NB-2, contactin 5, CNTN5, Neural recognition molecule NB-2
Species Reactivity Mouse
Antigen Contactin 5
Host Species Rabbit
Immunogen A synthetic peptide from mouse Contactin-5
Quantity 100μL
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled

Contactin 5 Antibody, OSC00019W, from Invitrogen. Species Reactivity: Mouse; Applications: Immunohistochemistry, Western Blot.


Host Species: Rabbit
Cross Reactivity: Mouse
Immunogen: A synthetic peptide from mouse Contactin-5
Formulation: 100μL unpurified antibody in whole serum and no preservative
Synonyms: NB-2, contactin 5, CNTN5, Neural recognition molecule NB-2
Uniprot Accession: P68500
Entrez GeneID: 244682
Gene Symbol: CNTN5
Reconstitute with 100 ul of distilled water.