Please login to your online account to display your discounted pricing

Contactin 4 Polyclonal Antibody, Invitrogen™


Contactin 4 Antibody, OSC00020W, from Invitrogen™. Species Reactivity: Rat; Applications: Immunohistochemistry, Western Blot.

Manufacturer: pierce biotechnology inc  OSC00020W

Catalog No. PIOSC00020W

  • / Each
 

Description & Specifications

Specifications

Antigen Contactin 4
Applications Immunohistochemistry
Applications Western Blot
Conjugate Unlabeled
Format Serum
Formulation 100μL unpurified antibody in whole serum and no preservative
Gene Accession No. Q62845
Gene Alias Brain-derived immunoglobulin superfamily protein 2, AXCAM, contactin 4, BIG-2, CNTN4A, contactin-4
Gene Symbols CNTN4
Host Species Rabbit
Immunogen A synthetic peptide from rat contactin-4
Quantity 100μL
Storage Requirements avoiding freeze/thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 116658
Target Species Rat

Contactin 4 Antibody, OSC00020W, from Invitrogen. Species Reactivity: Rat; Applications: Immunohistochemistry, Western Blot.


Host Species: Rabbit
Cross Reactivity: Rat
Immunogen: A synthetic peptide from rat contactin-4
Formulation: 100μL unpurified antibody in whole serum and no preservative
Synonyms: Brain-derived immunoglobulin superfamily protein 2, AXCAM, contactin 4, BIG-2, CNTN4A, contactin-4
Uniprot Accession: Q62845
Entrez GeneID: 116658
Gene Symbol: CNTN4
Reconstitute with 100 ul of distilled water.