Please login to your online account to display your discounted pricing

anti-Contactin 4 Polyclonal, Thermo Scientific™


Contactin 4 Polyclonal antibody OSC00014W from Thermo Scientific™ specifically detects Contactin 4 in Human samples, and it is validated for Immunohistochemistry and Western Blot

Manufacturer: pierce biotechnology inc  OSC00018W

 View more versions of this product

Catalog No. PIOSC00018W

  • / Each
 

Description & Specifications

Specifications

Formulation 100μL unpurified antibody in whole serum and no preservative
Gene Symbols CNTN4
Applications Immunohistochemistry
Applications Western Blot
Gene ID (Entrez) 269784
Cross Reactivity Mouse
Storage Requirements avoiding freeze/thaw cycles.
Format Serum
Gene Accession No. Q69Z26
Gene Alias Brain-derived immunoglobulin superfamily protein 2, Kiaa3024, contactin 4, BIG-2, SCA16, CNTN4A
Species Reactivity Mouse
Quantity 100μL
Primary or Secondary Primary
Conjugate Unlabeled
Antigen Contactin 4
Host Species Rabbit
Immunogen A synthetic peptide from mouse contactin-4
Monoclonal or Polyclonal Polyclonal

Contactin 4 Polyclonal antibody OSC00014W from Thermo Scientific™ specifically detects Contactin 4 in Human samples, and it is validated for Immunohistochemistry and Western Blot


Host Species: Rabbit
Cross Reactivity: Mouse
Immunogen: A synthetic peptide from mouse contactin-4
Formulation: 100μL unpurified antibody in whole serum and no preservative
Synonyms: Brain-derived immunoglobulin superfamily protein 2, Kiaa3024, contactin 4, BIG-2, SCA16, CNTN4A
Uniprot Accession: Q69Z26
Entrez GeneID: 269784
Gene Symbol: CNTN4
Reconstitute with 100 ul of distilled water.