Please login to your online account to display your discounted pricing

Contactin 4 Polyclonal Antibody, Invitrogen™


Contactin 4 Antibody, OSC00014W, from Invitrogen™. Species Reactivity: Human; Applications: Immunohistochemistry, Western Blot.

Manufacturer: pierce biotechnology inc  OSC00014W

Catalog No. PIOSC00014W

  • / Each
 

Description & Specifications

Specifications

Antigen Contactin 4
Applications Immunohistochemistry
Applications Western Blot
Conjugate Unlabeled
Format Serum
Formulation 100μL unpurified antibody in whole serum and no preservative
Gene Accession No. Q8IWV2
Gene Alias Brain-derived immunoglobulin superfamily protein 2, AXCAM, contactin 4, BIG-2, SCA16, CNTN4A, MGC33615
Gene Symbols CNTN4
Host Species Rabbit
Immunogen A synthetic peptide from human Contactin-4
Quantity 100μL
Storage Requirements avoiding freeze/thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 152330
Target Species Human

Contactin 4 Antibody, OSC00014W, from Invitrogen. Species Reactivity: Human; Applications: Immunohistochemistry, Western Blot.


Host Species: Rabbit
Cross Reactivity: Human
Immunogen: A synthetic peptide from human Contactin-4
Formulation: 100μL unpurified antibody in whole serum and no preservative
Synonyms: Brain-derived immunoglobulin superfamily protein 2, AXCAM, contactin 4, BIG-2, SCA16, CNTN4A, MGC33615
Uniprot Accession: Q8IWV2
Entrez GeneID: 152330
Gene Symbol: CNTN4
Reconstitute with 100 ul of distilled water.