Please login to your online account to display your discounted pricing

anti-Contactin 4 Polyclonal, Thermo Scientific™

Contactin 4 Polyclonal antibody OSC00014W from Thermo Scientific™ specifically detects Contactin 4 in Human samples, and it is validated for Immunohistochemistry and Western Blot

$448.05 - $448.05

Specifications

Antigen Contactin 4
Cross Reactivity Human
Format Serum
Host Species Rabbit
Immunogen A synthetic peptide from human Contactin-4
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

PIOSC00018W

 
pierce biotechnology inc
OSC00018W
Unlabeled 100μL Each for $448.05

PIOSC00014W

 
pierce biotechnology inc
OSC00014W
Unlabeled 100μL Each for $448.05

PIOSC00020W

 
pierce biotechnology inc
OSC00020W
Unlabeled 100μL Each for $448.05
Description & Specifications

Specifications

Antigen Contactin 4
Cross Reactivity Human
Format Serum
Host Species Rabbit
Immunogen A synthetic peptide from human Contactin-4
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Storage Requirements avoiding freeze/thaw cycles.

Contactin 4 Polyclonal antibody OSC00014W from Thermo Scientific™ specifically detects Contactin 4 in Human samples, and it is validated for Immunohistochemistry and Western Blot


Host Species: Rabbit
Cross Reactivity: Human
Immunogen: A synthetic peptide from human Contactin-4
Formulation: 100μL unpurified Antibody in whole serum, with no preservative;
Synonyms: Brain-derived immunoglobulin superfamily protein 2, AXCAM, contactin 4, BIG-2, SCA16, CNTN4A, MGC33615
Uniprot Accession: Q8IWV2
Gene Symbol: CNTN4
Entrez GeneID: 152330
OSC00014W detects Contactin 4 in Human samples. OSC00014W has been successfully used in Immunohistochemistry and Western Blot procedures. OSC00014W The immunogen is A synthetic peptide from human Contactin-4.