Please login to your online account to display your discounted pricing

Contactin 2 Polyclonal Antibody, Invitrogen™


Contactin 2 Antibody, OSC00022W, from Invitrogen™. Species Reactivity: Human; Applications: Immunohistochemistry, Western Blot.

Manufacturer: pierce biotechnology inc  OSC00022W

Catalog No. PIOSC00022W

  • / Each
 

Description & Specifications

Specifications

Antigen Contactin 2
Applications Immunohistochemistry
Applications Western Blot
Conjugate Unlabeled
Format Serum
Formulation 100μL unpurified antibody in whole serum and no preservative
Gene Accession No. Q02246
Gene Alias Axonin-1, Axonal glycoprotein TAG-1, Transient axonal glycoprotein 1, TAX-1, CNTN2, AXT, TAG1, TAX1, MGC157722, DKFZp781D102
Gene Symbols CNTN2
Host Species Rabbit
Immunogen A synthetic peptide from human contactin-2
Quantity 100μL
Storage Requirements avoiding freeze/thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 6900
Target Species Human

Contactin 2 Antibody, OSC00022W, from Invitrogen. Species Reactivity: Human; Applications: Immunohistochemistry, Western Blot.


Host Species: Rabbit
Cross Reactivity: Human
Immunogen: A synthetic peptide from human contactin-2
Formulation: 100μL unpurified antibody in whole serum and no preservative
Synonyms: Axonin-1, Axonal glycoprotein TAG-1, Transient axonal glycoprotein 1, TAX-1, CNTN2, AXT, TAG1, TAX1, MGC157722, DKFZp781D102
Uniprot Accession: Q02246
Entrez GeneID: 6900
Gene Symbol: CNTN2
Reconstitute with 100 ul of distilled water.