Please login to your online account to display your discounted pricing

anti-Contactin 2 Polyclonal, Thermo Scientific™

Contactin 2 Polyclonal antibody OSC00022W from Thermo Scientific™ specifically detects Contactin 2 in Human samples, and it is validated for Immunohistochemistry and Western Blot

$448.05 - $448.05

Specifications

Antigen Contactin 2
Cross Reactivity Mouse
Cross Reactivity Rat
Format Serum
Host Species Rabbit
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

PIOSC00022W

 
pierce biotechnology inc
OSC00022W
Unlabeled 100μL Each for $448.05

PIOSC00012W

 
pierce biotechnology inc
OSC00012W
Unlabeled 100μL Each for $448.05
Description & Specifications

Specifications

Antigen Contactin 2
Cross Reactivity Mouse
Cross Reactivity Rat
Format Serum
Host Species Rabbit
Immunogen A synthetic peptide from rat contactin-2
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Storage Requirements avoiding freeze/thaw cycles.

Contactin 2 Polyclonal antibody OSC00022W from Thermo Scientific™ specifically detects Contactin 2 in Human samples, and it is validated for Immunohistochemistry and Western Blot


Host Species: Rabbit
Cross Reactivity: Human
Immunogen: A synthetic peptide from human contactin-2
Formulation: 100μL unpurified Antibody in whole serum, with no preservative;
Synonyms: Axonin-1, Axonal glycoprotein TAG-1, Transient axonal glycoprotein 1, TAX-1, CNTN2, AXT, TAG1, TAX1, MGC157722, DKFZp781D102
Uniprot Accession: Q02246
Gene Symbol: CNTN2
Entrez GeneID: 6900
OSC00022W detects Contactin 2 in Human samples. OSC00022W has been successfully used in Immunohistochemistry and Western Blot procedures. OSC00022W The immunogen is A synthetic peptide from human contactin-2.