Please login to your online account to display your discounted pricing

Contactin 2 Polyclonal Antibody, Invitrogen™


Contactin 2 Antibody, OSC00012W, from Invitrogen™. Species Reactivity: Mouse, Rat; Applications: Immunohistochemistry, Western Blot.

Manufacturer: pierce biotechnology inc  OSC00012W

Catalog No. PIOSC00012W

  • / Each
 

Description & Specifications

Specifications

Applications Immunohistochemistry
Applications Western Blot
Cross Reactivity Mouse
Cross Reactivity Rat
Gene Symbols CNTN2
Formulation 100μL unpurified antibody in whole serum and no preservative
Gene ID (Entrez) 21367, 25356
Storage Requirements avoiding freeze/thaw cycles.
Gene Accession No. P22063, Q61330
Format Serum
Species Reactivity Mouse, Rat
Gene Alias Axonin-1, Axonal glycoprotein TAG-1, Transient axonal glycoprotein 1, TAX-1, CNTN2, AXT, TAG1, TAX1, MGC157722, DKFZp781D102
Antigen Contactin 2
Host Species Rabbit
Immunogen A synthetic peptide from rat contactin-2
Quantity 100μL
Conjugate Unlabeled
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal

Contactin 2 Antibody, OSC00012W, from Invitrogen. Species Reactivity: Mouse, Rat; Applications: Immunohistochemistry, Western Blot.


Host Species: Rabbit
Cross Reactivity: Mouse, Rat
Immunogen: A synthetic peptide from rat contactin-2
Formulation: 100μL unpurified antibody in whole serum and no preservative
Synonyms: Axonin-1, Axonal glycoprotein TAG-1, Transient axonal glycoprotein 1, TAX-1, CNTN2, AXT, TAG1, TAX1, MGC157722, DKFZp781D102
Uniprot Accession: P22063, Q61330
Entrez GeneID: 21367, 25356
Gene Symbol: CNTN2
Reconstitute with 100 ul of distilled water.