Please login to your online account to display your discounted pricing

anti-Contactin 1 Polyclonal, Thermo Scientific™


Contactin 1 Polyclonal Ab OSC00027W from Thermo Scientific™ specifically detects Contactin 1 in Hu and Rt samples, and it is validated for IHC and WB

Manufacturer: pierce biotechnology inc  OSC00026W

 View more versions of this product

Catalog No. PIOSC00026W

  • / Each
 

Description & Specifications

Specifications

Formulation 100μL unpurified antibody in whole serum and no preservative
Applications Immunohistochemistry
Applications Western Blot
Gene ID (Entrez) 12805
Cross Reactivity Mouse
Gene Symbols CNTN1
Gene Accession No. P12960
Gene Alias Neural cell surface protein F3, F3cam, contactin 1, CNTN1
Species Reactivity Mouse
Storage Requirements avoiding freeze/thaw cycles.
Format Serum
Antigen Contactin 1
Host Species Rabbit
Quantity 100μL
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled
Immunogen A synthetic peptide from mouse Contactin-1

Contactin 1 Polyclonal Ab OSC00027W from Thermo Scientific™ specifically detects Contactin 1 in Hu and Rt samples, and it is validated for IHC and WB


Host Species: Rabbit
Cross Reactivity: Mouse
Immunogen: A synthetic peptide from mouse Contactin-1
Formulation: 100μL unpurified antibody in whole serum and no preservative
Synonyms: Neural cell surface protein F3, F3cam, contactin 1, CNTN1
Uniprot Accession: P12960
Entrez GeneID: 12805
Gene Symbol: CNTN1
Reconstitute with 100 ul of distilled water.