Please login to your online account to display your discounted pricing

anti-Contactin 1 Polyclonal, Thermo Scientific™

Contactin 1 Polyclonal Ab OSC00027W from Thermo Scientific™ specifically detects Contactin 1 in Hu and Rt samples, and it is validated for IHC and WB

$448.05 - $448.05

Specifications

Antigen Contactin 1
Cross Reactivity Human
Cross Reactivity Rat
Format Serum
Host Species Rabbit
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

PIOSC00026W

 
pierce biotechnology inc
OSC00026W
Unlabeled 100μL Each for $448.05

PIOSC00027W

 
pierce biotechnology inc
OSC00027W
Unlabeled 100μL Each for $448.05
Description & Specifications

Specifications

Antigen Contactin 1
Cross Reactivity Human
Cross Reactivity Rat
Format Serum
Host Species Rabbit
Immunogen A synthetic peptide from human contactin-1
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Storage Requirements avoiding freeze/thaw cycles.

Contactin 1 Polyclonal Ab OSC00027W from Thermo Scientific™ specifically detects Contactin 1 in Hu and Rt samples, and it is validated for IHC and WB


Host Species: Rabbit
Cross Reactivity: Human, Rat
Immunogen: A synthetic peptide from human contactin-1
Formulation: 100μL unpurified Antibody in whole serum, with no preservative;
Synonyms: Neural cell surface protein F3, F3cam, contactin 1, CNTN1, Glycoprotein gp135
Uniprot Accession: Q12860, Q63198
Gene Symbol: CNTN1
Entrez GeneID: 117258, 1272
OSC00027W detects Contactin 1 in Human and Rat samples. OSC00027W has been successfully used in Immunohistochemistry and Western Blot procedures. OSC00027W The immunogen is A synthetic peptide from human contactin-1.