Please login to your online account to display your discounted pricing

Contactin 1 Polyclonal Antibody, Invitrogen™


Contactin 1 Antibody, OSC00027W, from Invitrogen™. Species Reactivity: Human, Rat; Applications: Immunohistochemistry, Western Blot.

Manufacturer: pierce biotechnology inc  OSC00027W

Catalog No. PIOSC00027W

  • / Each
 

Description & Specifications

Specifications

Formulation 100μL unpurified antibody in whole serum and no preservative
Applications Immunohistochemistry
Applications Western Blot
Cross Reactivity Human
Cross Reactivity Rat
Storage Requirements avoiding freeze/thaw cycles.
Gene Symbols CNTN1
Gene Alias Neural cell surface protein F3, F3cam, contactin 1, CNTN1, Glycoprotein gp135
Species Reactivity Human, Rat
Gene ID (Entrez) 117258, 1272
Format Serum
Gene Accession No. Q12860, Q63198
Quantity 100μL
Immunogen A synthetic peptide from human contactin-1
Conjugate Unlabeled
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Antigen Contactin 1
Host Species Rabbit

Contactin 1 Antibody, OSC00027W, from Invitrogen. Species Reactivity: Human, Rat; Applications: Immunohistochemistry, Western Blot.


Host Species: Rabbit
Cross Reactivity: Human, Rat
Immunogen: A synthetic peptide from human contactin-1
Formulation: 100μL unpurified antibody in whole serum and no preservative
Synonyms: Neural cell surface protein F3, F3cam, contactin 1, CNTN1, Glycoprotein gp135
Uniprot Accession: Q12860, Q63198
Entrez GeneID: 117258, 1272
Gene Symbol: CNTN1
Reconstitute with 100 ul of distilled water.