Please login to your online account to display your discounted pricing

anti-Collagen I alpha 1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP177458

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP177458

  • / Each
 

Description & Specifications

Specifications

Test Specificity Mouse. Predicted reactivity in Human.
Immunogen A synthetic peptide made to the C-terminal portion of the human Collagen I protein (between residues 1300-1350) [UniProt P02452]
Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled
Formulation PBS, 30% glycerol
Gene ID (Entrez) 1277
Concentration 1.12mg/mL
Antigen Collagen I alpha 1
Gene Accession No. P02452
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Format Affinity Purified
Host Species Rabbit
Quantity 0.1mL
Cross Reactivity Human
Cross Reactivity Mouse
Primary or Secondary Primary
Regulatory Status RUO