Please login to your online account to display your discounted pricing

anti-Collagen I alpha 1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP177457

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP177457

  • / Each
 

Description & Specifications

Specifications

Antigen Collagen I alpha 1
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Concentration 1.1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Format Affinity Purified
Formulation PBS, 30% glycerol
Gene Accession No. P02452
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human Collagen I protein (between residues 150-200) [UniProt P02452]
Quantity 0.1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 1277
Test Specificity Mouse. Predicted reactivity in Human.
Target Species Human
Target Species Murine