Please login to your online account to display your discounted pricing

anti-Collagen I alpha 1, Biotin, Polyclonal, Novus Biologicals™

$289.00 - $406.00

Specifications

Host Species Rabbit
Isotype IgG
Immunogen Collagen Type I from adult human and bovine tissues.
Antigen Collagen I alpha 1
Monoclonal or Polyclonal Polyclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP177458B

 
novus biologicals canada
NBP177458B
Biotin 0.1mL Each for $406.00

NB1206577

 
novus biologicals canada
NB1206577
Biotin 0.05mg Each for $289.00
Description & Specifications

Specifications

Host Species Rabbit
Isotype IgG
Immunogen Collagen Type I from adult human and bovine tissues.
Antigen Collagen I alpha 1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Test Specificity Negligible cross-reactivity with Type II, III, IV, V or VI collagens. Non-specific cross reaction of anti-collagen antibodies with other human serum proteins or non-collagen extracellular matrix proteins is negligible. Cross-reacts with Human and Cow. Expected to cross-react with all Mammals due to sequence identity. Not yet tested in other species.
Gene Accession No. P02452
Gene ID (Entrez) 1277
Formulation 0.1M Sodium Borate, 0.075M Sodium Chloride, 0.005M EDTA, pH 8.0 with 10mg/mL BSA
Concentration 1.0mg/mL