Please login to your online account to display your discounted pricing

anti-Collagen I alpha 1, Biotin, Polyclonal, Novus Biologicals™

Manufacturer: novus biologicals canada  NB1206577

 View all options for this product


For Research Use Only

Catalog No. NB1206577


Description & Specifications


Antigen Collagen I alpha 1
Applications ELISA
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Concentration 1.0mg/mL
Conjugate Biotin
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Affinity Purified
Formulation 0.1M Sodium Borate, 0.075M Sodium Chloride, 0.005M EDTA, pH 8.0 with 10mg/mL BSA
Gene Accession No. P02452
Host Species Rabbit
Immunogen Collagen Type I from adult human and bovine tissues.
Isotype IgG
Quantity 0.05mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 1277
Test Specificity Negligible cross-reactivity with Type II, III, IV, V or VI collagens. Non-specific cross reaction of anti-collagen antibodies with other human serum proteins or non-collagen extracellular matrix proteins is negligible. Cross-reacts with Human and Cow. Expected to cross-react with all Mammals due to sequence identity. Not yet tested in other species.