Please login to your online account to display your discounted pricing

Coagulation Factor III/Tissue Factor Mouse anti-Human, Clone: 323514, R&D Systems

Mouse Monoclonal Antibody

$317.73 - $317.73

Specifications

Antigen Coagulation Factor III/Tissue Factor
Clone 323514
Cross Reactivity Human
Format Purified
Host Species Mouse
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

MAB2339100

 
r & d systems
RDSMAB2339100
Unlabeled 100μg Each for $317.73
Description & Specifications

Specifications

Antigen Coagulation Factor III/Tissue Factor
Clone 323514
Cross Reactivity Human
Format Purified
Host Species Mouse
Regulatory Status RUO
Immunogen Mouse myeloma cell line NS0-derived recombinant human Coagulation Factor III. Gly34-Glu251
Isotype IgG2a
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Purification Method Protein A or G purified from hybridoma culture supernatant

Coagulation Factor III/Tissue Factor Monoclonal antibody MAB2339 Specifically detects Coagulation Factor III/Tissue Factor in Human samples. It is validated for ELISA Capture, Immunoprecipitation, Western Blotting