Please login to your online account to display your discounted pricing

anti-CNTF Polyclonal, Thermo Scientific™

CNTF Polyclonal antibody PA1-18358 from Thermo Scientific™ specifically detects CNTF in Human, Mouse and Rat samples, and it is validated for Western Blot

$494.55 - $509.25

Specifications

Antigen CNTF
Cross Reactivity Human
Cross Reactivity Rat
Format Serum
Host Species Sheep
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

01-670-610

 
pierce biotechnology inc
PA118358
Unlabeled 100μL Each for $494.55

01-670-617

 
pierce biotechnology inc
PA118365
Unlabeled 100μL Each for $509.25
Description & Specifications

Specifications

Antigen CNTF
Cross Reactivity Human
Cross Reactivity Rat
Format Serum
Host Species Sheep
Immunogen Recombinant human CNTF.
Isotype IgG
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Storage Requirements Avoid Freeze/Thaw Cycles

CNTF Polyclonal antibody PA1-18358 from Thermo Scientific™ specifically detects CNTF in Human, Mouse and Rat samples, and it is validated for Western Blot

CNTF is a survival promoting factor for different types of neurons in vitro and in vivo. The essential structural features for the biological function of human CNTF were investigated by Thier, M. et al. They showed that deletion of 14 N-terminal and 18 C-terminal amino acids significantly increased bioactivity compared to wild-type CNTF. CNTF is a survival factor for various neuronal cell types, which seems to prevent the degeneration of motor axons after axotomy.
Host Species: Rabbit
Cross Reactivity: Human, Mouse, Rat
Isotype: IgG
Immunogen: Recombinant human CNTF.
Formulation: 100μL unpurified Antibody in whole serum, with no preservative;
Synonyms: Ciliary neurotrophic factor
Uniprot Accession: P20294, P26441, P51642
Gene Symbol: CNTF
Entrez GeneID: 1270, 12803, 25707
PA1-18358 detects CNTF in Human, Mouse and Rat samples. PA1-18358 has been successfully used in Western Blot procedures. PA1-18358 The immunogen is Recombinant human CNTF..