Please login to your online account to display your discounted pricing

anti-CLOCK, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP130326

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP130326

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Polyclonal
Host Species Rabbit
Cross Reactivity Human
Cross Reactivity Mouse
Concentration 1mg/mL
Formulation PBS, 30% glycerol
Gene ID (Entrez) 9575
Test Specificity Human and mouse. Immunogen has 94% identity with rat. Other species have not been tested.
Conjugate Unlabeled
Applications Immunohistochemistry
Applications Western Blotting
Format Affinity Purified
Immunogen Synthetic peptide made to a C-terminal portion of mouse CLOCK (within residues 805-855). [Swiss-Prot No. O08785]
Antigen CLOCK
Primary or Secondary Primary
Quantity 0.1mL
Regulatory Status RUO