Please login to your online account to display your discounted pricing

anti-CLOCK, Polyclonal, Novus Biologicals™

$106.00 - $321.00

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to a C-terminal portion of mouse CLOCK (within residues 805-855). [Swiss-Prot No. O08785]
Antigen CLOCK
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP130326

 
novus biologicals canada
NBP130326
Unlabeled 0.1mL Each for $321.00

NBP130326SS

 
novus biologicals canada
NBP130326SS
Unlabeled 0.025mL Each for $106.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to a C-terminal portion of mouse CLOCK (within residues 805-855). [Swiss-Prot No. O08785]
Antigen CLOCK
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Western Blotting
Test Specificity Human and mouse. Immunogen has 94% identity with rat. Other species have not been tested.
Gene ID (Entrez) 9575
Formulation PBS, 30% glycerol
Concentration 1mg/mL