Please login to your online account to display your discounted pricing

anti-CLOCK, Polyclonal, Novus Biologicals™

$139.00 - $309.05

Specifications

Host Species Rabbit
Immunogen Two peptides derived from mouse CLOCK conjugated to albumin. [UniProt No. O08785]
Antigen CLOCK
Monoclonal or Polyclonal Polyclonal
Format Antisera
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB100126

 
novus biologicals canada
NB100126
Unlabeled 0.1mL Each for $309.05

NB100126SS

 
novus biologicals canada
NB100126SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Two peptides derived from mouse CLOCK conjugated to albumin. [UniProt No. O08785]
Antigen CLOCK
Monoclonal or Polyclonal Polyclonal
Format Antisera
Primary or Secondary Primary
Regulatory Status RUO
Applications Chromatin Immunoprecipitation
Applications Western Blot
Gene ID (Entrez) 9575