Please login to your online account to display your discounted pricing

anti-CLOCK, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP130326SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP130326SS

  • / Each
 

Description & Specifications

Specifications

Antigen CLOCK
Applications Immunohistochemistry
Applications Western Blotting
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Format Affinity Purified
Formulation PBS, 30% glycerol
Host Species Rabbit
Immunogen Synthetic peptide made to a C-terminal portion of mouse CLOCK (within residues 805-855). [Swiss-Prot No. O08785]
Quantity 0.025mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 9575
Test Specificity Human and mouse. Immunogen has 94% identity with rat. Other species have not been tested.