Please login to your online account to display your discounted pricing

anti-CLEC4E, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Immunogen This antibody was raised against CLEC4E
Antigen CLEC4E
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP227093

 
novus biologicals canada
NBP227093
Unlabeled 0.1mg Each for $255.80

NBP227093SS

 
novus biologicals canada
NBP227093SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen This antibody was raised against CLEC4E
Antigen CLEC4E
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen for this antibody is 100% homologous in human, common chimpanzee, orangutan, and white-cheeked gibbon, 94% in crab-eating macaque, rhesus monkey, and sheep, 88% in cow, rat, and new world monkey, 82% in panda, dog, horse, and african bush elephant, 76% in guinea pig, and naked mole rat, 70% in mouse, and rabbit, 64% in Chinese hamster, opossum, and platypus.
Gene Accession No. Q9ULY5
Gene ID (Entrez) 26253
Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA.
Concentration 0.5mg/mL