Please login to your online account to display your discounted pricing

anti-CLEC-1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP222382SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP222382SS

  • / Each
 

Description & Specifications

Specifications

Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA
Conjugate Unlabeled
Host Species Mouse
Gene Accession No. NP_057595.2
Gene ID (Entrez) 51267
Test Specificity Chimpanzee, Human, Orangutan, White-cheeked gibbon. The amino acid sequence used as immunogen for this antibody is 100% homologous in human, orangutan, chimpanzee, and white-cheeked gibbon, 89% in new world monkey, crab-eating macaque, and rhesus monkey, 78% in pig, 72% in dog, african bush elephant, and rabbit, 67% in panda, cattle, Chinese hamster, and horse, 61% in rat.
Quantity 0.025mg
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Monoclonal or Polyclonal Polyclonal
Regulatory Status RUO
Antigen CLEC-1
Isotype κ
Cross Reactivity Human
Cross Reactivity Primate
Primary or Secondary Primary
Format Purified
Immunogen A portion of amino acids 50-100 from human CLEC1 was used as the immunogen for this antibody.