Please login to your online account to display your discounted pricing

anti-CLEC-1, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Mouse
Isotype κ
Immunogen A portion of amino acids 50-100 from human CLEC1 was used as the immunogen for this antibody.
Antigen CLEC-1
Monoclonal or Polyclonal Polyclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP222382SS

 
novus biologicals canada
NBP222382SS
Unlabeled 0.025mg Each for $139.00

NBP222382

 
novus biologicals canada
NBP222382
Unlabeled 0.1mg Each for $255.80
Description & Specifications

Specifications

Host Species Mouse
Isotype κ
Immunogen A portion of amino acids 50-100 from human CLEC1 was used as the immunogen for this antibody.
Antigen CLEC-1
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Test Specificity Chimpanzee, Human, Orangutan, White-cheeked gibbon. The amino acid sequence used as immunogen for this antibody is 100% homologous in human, orangutan, chimpanzee, and white-cheeked gibbon, 89% in new world monkey, crab-eating macaque, and rhesus monkey, 78% in pig, 72% in dog, african bush elephant, and rabbit, 67% in panda, cattle, Chinese hamster, and horse, 61% in rat.
Gene Accession No. NP_057595.2
Gene ID (Entrez) 51267
Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA