Please login to your online account to display your discounted pricing

anti-CIDEB, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10055757SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10055757T

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Concentration 0.5mg/mL
Formulation PBS
Gene ID (Entrez) 27141
Test Specificity In mouse liver, an approx. 25 kDa band is observed.
Host Species Rabbit
Antigen CIDEB
Cross Reactivity Mouse
Conjugate Unlabeled
Immunogen The polyclonal antibody was raised against a peptide corresponding to amino acids 204 to 219 of murine CIDE-B.
Format Purified
Quantity 0.025mg
Regulatory Status RUO
Applications Western Blotting