Please login to your online account to display your discounted pricing

anti-CIDEB, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10055757

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10055757

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 27141
Formulation PBS
Conjugate Unlabeled
Primary or Secondary Primary
Immunogen The polyclonal antibody was raised against a peptide corresponding to amino acids 204 to 219 of murine CIDE-B.
Concentration 0.5mg/mL
Test Specificity In mouse liver, an approx. 25 kDa band is observed.
Antigen CIDEB
Format Purified
Quantity 0.1mg
Host Species Rabbit
Regulatory Status RUO
Cross Reactivity Mouse
Applications Western Blotting