Please login to your online account to display your discounted pricing

anti-CIDEB, Polyclonal, Novus Biologicals™

$139.00 - $309.05

Specifications

Host Species Rabbit
Immunogen The polyclonal antibody was raised against a peptide corresponding to amino acids 204 to 219 of murine CIDE-B.
Antigen CIDEB
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10055757

 
novus biologicals canada
NB10055757
Unlabeled 0.1mg Each for $309.05

NB10055757T

 
novus biologicals canada
NB10055757SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen The polyclonal antibody was raised against a peptide corresponding to amino acids 204 to 219 of murine CIDE-B.
Antigen CIDEB
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity In mouse liver, an approx. 25 kDa band is observed.
Gene ID (Entrez) 27141
Formulation PBS
Concentration 0.5mg/mL