Please login to your online account to display your discounted pricing

anti-cIAP-2/HIAP-1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP127972SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP127972SS

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Gene Accession No. Q13489
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Simple Western
Applications Western Blotting
Concentration 1.5mg/mL
Gene ID (Entrez) 330
Formulation PBS, 30% glycerol
Conjugate Unlabeled
Host Species Rabbit
Antigen cIAP-2/HIAP-1
Quantity 0.025mL
Cross Reactivity Human
Regulatory Status RUO
Immunogen Synthetic peptide made to an internal portion of human cIAP2 (within residues 100-200). [Swiss-Prot No. Q13489]
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified