Please login to your online account to display your discounted pricing

anti-cIAP-2/HIAP-1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP127972

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP127972

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 330
Formulation PBS, 30% glycerol
Primary or Secondary Primary
Quantity 0.1mL
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Simple Western
Applications Western Blotting
Conjugate Unlabeled
Concentration 1.5mg/mL
Gene Accession No. Q13489
Regulatory Status RUO
Format Affinity Purified
Immunogen Synthetic peptide made to an internal portion of human cIAP2 (within residues 100-200). [Swiss-Prot No. Q13489]
Cross Reactivity Human
Antigen cIAP-2/HIAP-1
Host Species Rabbit