Please login to your online account to display your discounted pricing

anti-cIAP-2/HIAP-1, Polyclonal, Novus Biologicals™

$144.63 - $222.95

Specifications

Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 520-570 (KYIPTEDVSDLPVEEQLR) of human cIAP2 was used as immunogen
Antigen cIAP-2/HIAP-1
Monoclonal or Polyclonal Polyclonal
Format Antisera
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056131T

 
novus biologicals canada
NB10056131SS
Unlabeled 0.025mL Each for $144.63

NB10056131

 
novus biologicals canada
NB10056131
Unlabeled 0.05mL Each for $222.95
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 520-570 (KYIPTEDVSDLPVEEQLR) of human cIAP2 was used as immunogen
Antigen cIAP-2/HIAP-1
Monoclonal or Polyclonal Polyclonal
Format Antisera
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Test Specificity cIAP2 . Reacts with Human. 88% sequence identity with bovine and 83% with Chicken, Porcine, Rat and mouse proteins.
Gene ID (Entrez) 330
Formulation PBS