Please login to your online account to display your discounted pricing

anti-cIAP-2/HIAP-1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056131SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056131T

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 330
Primary or Secondary Primary
Test Specificity cIAP2 . Reacts with Human. 88% sequence identity with bovine and 83% with Chicken, Porcine, Rat and mouse proteins.
Conjugate Unlabeled
Formulation PBS
Immunogen A synthetic peptide corresponding to amino acids 520-570 (KYIPTEDVSDLPVEEQLR) of human cIAP2 was used as immunogen
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Format Antiserum
Cross Reactivity Human
Regulatory Status RUO
Quantity 0.025mL
Antigen cIAP-2/HIAP-1
Monoclonal or Polyclonal Polyclonal
Host Species Rabbit