Please login to your online account to display your discounted pricing

anti-cIAP-2/HIAP-1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056131

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056131

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Formulation PBS
Gene ID (Entrez) 330
Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled
Test Specificity cIAP2 . Reacts with Human. 88% sequence identity with bovine and 83% with Chicken, Porcine, Rat and mouse proteins.
Immunogen A synthetic peptide corresponding to amino acids 520-570 (KYIPTEDVSDLPVEEQLR) of human cIAP2 was used as immunogen
Format Antiserum
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Regulatory Status RUO
Cross Reactivity Human
Quantity 0.05mL
Host Species Rabbit
Antigen cIAP-2/HIAP-1