Please login to your online account to display your discounted pricing

anti-cIAP-1/HIAP-2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227190SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227190SS

  • / Each
 

Description & Specifications

Specifications

Antigen cIAP-1/HIAP-2
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Conjugate Unlabeled
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Antisera
Formulation 50μL neat serum.
Gene Accession No. Q13490
Host Species Rabbit
Immunogen Full-length recombinant human cAIP1 was used as immunogen
Quantity 0.025mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 329
Target Species Canine
Target Species Human
Target Species Murine
Target Species Rat