Please login to your online account to display your discounted pricing

anti-cIAP-1/HIAP-2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227190

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227190

  • / Each
 

Description & Specifications

Specifications

Formulation 50μL neat serum.
Conjugate Unlabeled
Regulatory Status RUO
Gene ID (Entrez) 329
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Quantity 0.05mL
Gene Accession No. Q13490
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Host Species Rabbit
Antigen cIAP-1/HIAP-2
Immunogen Full-length recombinant human cAIP1 was used as immunogen
Format Antiserum