Please login to your online account to display your discounted pricing

anti-cIAP-1/HIAP-2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056130SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056130T

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Formulation PBS
Gene ID (Entrez) 329
Format Antiserum
Host Species Rabbit
Test Specificity cIAP1 has 618 amino acids and migrates at ~70 kDa on SDS-PAGE. Observed molecular weights may vary as genes generally produce, by alternative splicing, multiple transcripts all with introns, putatively encoding multiple different protein isoforms. Additionally, proteins may undergo post translational modifications including phosphorylation, glycosylation and cleavage which can result in differential molecular weights. Protein modifications can vary between cell and tissue type, and proteins can migrate as one or multiple bands depending on the isoform expressed as well as post-translational modifications.
Antigen cIAP-1/HIAP-2
Immunogen A synthetic peptide corresponding to amino acids 2-18 (HKTASQRLFPGPSYQNI) of human cIAP1 was used as immunogen
Quantity 0.025mL
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Regulatory Status RUO
Cross Reactivity Human