Please login to your online account to display your discounted pricing

anti-cIAP-1/HIAP-2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056130

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056130

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 329
Monoclonal or Polyclonal Polyclonal
Test Specificity cIAP1 has 618 amino acids and migrates at ~70 kDa on SDS-PAGE. Observed molecular weights may vary as genes generally produce, by alternative splicing, multiple transcripts all with introns, putatively encoding multiple different protein isoforms. Additionally, proteins may undergo post translational modifications including phosphorylation, glycosylation and cleavage which can result in differential molecular weights. Protein modifications can vary between cell and tissue type, and proteins can migrate as one or multiple bands depending on the isoform expressed as well as post-translational modifications.
Primary or Secondary Primary
Formulation PBS
Format Antiserum
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Regulatory Status RUO
Quantity 0.05mL
Cross Reactivity Human
Immunogen A synthetic peptide corresponding to amino acids 2-18 (HKTASQRLFPGPSYQNI) of human cIAP1 was used as immunogen
Antigen cIAP-1/HIAP-2
Conjugate Unlabeled
Host Species Rabbit