Please login to your online account to display your discounted pricing

anti-cIAP-1/HIAP-2, Polyclonal, Novus Biologicals™

$160.00 - $255.80

Specifications

Host Species Rabbit
Immunogen Full-length recombinant human cAIP1 was used as immunogen
Antigen cIAP-1/HIAP-2
Monoclonal or Polyclonal Polyclonal
Format Antisera
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP227190

 
novus biologicals canada
NBP227190
Unlabeled 0.05mL Each for $255.80

NBP227190SS

 
novus biologicals canada
NBP227190SS
Unlabeled 0.025mL Each for $160.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Full-length recombinant human cAIP1 was used as immunogen
Antigen cIAP-1/HIAP-2
Monoclonal or Polyclonal Polyclonal
Format Antisera
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Gene Accession No. Q13490
Gene ID (Entrez) 329
Formulation 50μL neat serum.