Please login to your online account to display your discounted pricing

anti-cIAP-1/HIAP-2, Polyclonal, Novus Biologicals™

$144.63 - $222.95

Specifications

Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 2-18 (HKTASQRLFPGPSYQNI) of human cIAP1 was used as immunogen
Antigen cIAP-1/HIAP-2
Monoclonal or Polyclonal Polyclonal
Format Antisera
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056130T

 
novus biologicals canada
NB10056130SS
Unlabeled 0.025mL Each for $144.63

NB10056130

 
novus biologicals canada
NB10056130
Unlabeled 0.05mL Each for $222.95
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 2-18 (HKTASQRLFPGPSYQNI) of human cIAP1 was used as immunogen
Antigen cIAP-1/HIAP-2
Monoclonal or Polyclonal Polyclonal
Format Antisera
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Test Specificity cIAP1 has 618 amino acids and migrates at ~70 kDa on SDS-PAGE. Observed molecular weights may vary as genes generally produce, by alternative splicing, multiple transcripts all with introns, putatively encoding multiple different protein isoforms. Additionally, proteins may undergo post translational modifications including phosphorylation, glycosylation and cleavage which can result in differential molecular weights. Protein modifications can vary between cell and tissue type, and proteins can migrate as one or multiple bands depending on the isoform expressed as well as post-translational modifications.
Gene ID (Entrez) 329
Formulation PBS