Please login to your online account to display your discounted pricing

anti-CHORDC1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP178304

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP178304

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Polyclonal
Formulation PBS, 30% glycerol
Gene Accession No. Q9UHD1
Concentration 0.65mg/mL
Primary or Secondary Primary
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Gene ID (Entrez) 26973
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Quantity 0.1mL
Regulatory Status RUO
Host Species Rabbit
Antigen CHORDC1
Format Affinity Purified
Immunogen A synthetic peptide made to an internal portion of the human CHORDC1 protein (between residues 100-250) [UniProt Q9UHD1]