Please login to your online account to display your discounted pricing

anti-CHORDC1, Polyclonal, Novus Biologicals™

$109.00 - $309.05

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human CHORDC1 protein (between residues 100-250) [UniProt Q9UHD1]
Antigen CHORDC1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP178304

 
novus biologicals canada
NBP178304
Unlabeled 0.1mL Each for $309.05

NBP178304SS

 
novus biologicals canada
NBP178304SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human CHORDC1 protein (between residues 100-250) [UniProt Q9UHD1]
Antigen CHORDC1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Gene Accession No. Q9UHD1
Gene ID (Entrez) 26973
Formulation PBS, 30% glycerol
Concentration 0.65mg/mL