Please login to your online account to display your discounted pricing

anti-CHD7, Polyclonal, Novus Biologicals™

$139.00 - $329.00

Specifications

Host Species Rabbit
Immunogen A genomic peptide made to an N-terminal region of the human Chd7 protein (within residues 25-200). [Swiss-Prot Q9P2D1]
Antigen CHD7
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP177393SS

 
novus biologicals canada
NBP177393SS
Unlabeled 0.025mL Each for $139.00

NBP177393

 
novus biologicals canada
NBP177393
Unlabeled 0.1mL Each for $329.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A genomic peptide made to an N-terminal region of the human Chd7 protein (within residues 25-200). [Swiss-Prot Q9P2D1]
Antigen CHD7
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Test Specificity Human and mouse. Immunogen has 85% identity to chicken Chd7.
Gene Accession No. Q9P2D1
Gene ID (Entrez) 55636
Formulation PBS, 30% glycerol
Concentration 1.27mg/mL