Please login to your online account to display your discounted pricing

anti-ch-TOG, Polyclonal, Novus Biologicals™

$139.00 - $362.00

Specifications

Host Species Rabbit
Immunogen A recombinant segment of the C-terminal domain of human ch-TOG [UniProt No. Q14008]
Antigen ch-TOG
Monoclonal or Polyclonal Polyclonal
Format Antisera
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB500182

 
novus biologicals canada
NB500182
Unlabeled 0.1mL Each for $362.00

NB500182SS

 
novus biologicals canada
NB500182SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A recombinant segment of the C-terminal domain of human ch-TOG [UniProt No. Q14008]
Antigen ch-TOG
Monoclonal or Polyclonal Polyclonal
Format Antisera
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blot
Gene Accession No. Q14008
Gene ID (Entrez) 9793