Please login to your online account to display your discounted pricing

anti-CENTB1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224631

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224631

  • / Each
 

Description & Specifications

Specifications

Formulation 100μg in 200μL PBS containing 0.05% BSA
Gene Accession No. Q15027
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoform CRA_a), chimpanzee and monkey, and 73% homologous in dog. Based on amino acid content, the theoretical size of TSLP protein is approximately 17 kD. However, it has been experimentally observedat approximately 20-30 kD.
Cross Reactivity Human
Conjugate Unlabeled
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 9744
Concentration 0.5mg/mL
Host Species Rabbit
Quantity 0.1mg
Format Purified
Antigen CENTB1
Immunogen A portion of amino acids 520-560 of human ACAP1 was used as the immunogen.
Regulatory Status RUO
Applications Western Blotting