Please login to your online account to display your discounted pricing

anti-CENTB1, Polyclonal, Novus Biologicals™

$109.00 - $257.41

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 520-560 of human ACAP1 was used as the immunogen.
Antigen CENTB1
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224631SS

 
novus biologicals canada
NBP224631SS
Unlabeled 0.025mg Each for $109.00

NBP224631

 
novus biologicals canada
NBP224631
Unlabeled 0.1mg Each for $257.41
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 520-560 of human ACAP1 was used as the immunogen.
Antigen CENTB1
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoform CRA_a), chimpanzee and monkey, and 73% homologous in dog. Based on amino acid content, the theoretical size of TSLP protein is approximately 17 kD. However, it has been experimentally observedat approximately 20-30 kD.
Gene Accession No. Q15027
Gene ID (Entrez) 9744
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL