Please login to your online account to display your discounted pricing

anti-CENTB1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224631SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224631SS

  • / Each
 

Description & Specifications

Specifications

Formulation 100μg in 200μL PBS containing 0.05% BSA
Gene Accession No. Q15027
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 9744
Format Purified
Applications Western Blotting
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoform CRA_a), chimpanzee and monkey, and 73% homologous in dog. Based on amino acid content, the theoretical size of TSLP protein is approximately 17 kD. However, it has been experimentally observedat approximately 20-30 kD.
Concentration 0.5mg/mL
Quantity 0.025mg
Conjugate Unlabeled
Immunogen A portion of amino acids 520-560 of human ACAP1 was used as the immunogen.
Regulatory Status RUO
Cross Reactivity Human
Antigen CENTB1
Host Species Rabbit