Please login to your online account to display your discounted pricing

anti-CENPF Polyclonal, Thermo Scientific™

CENPF Polyclonal Ab PA1-16826 from Thermo Scientific™ specifically detects CENPF in Bv and Hu samples, and it is validated for Flow Cytometry, ICC, IF, IP and WB

$448.05 - $448.05

Specifications

Antigen CENPF
Cross Reactivity Bovine
Cross Reactivity Human
Format Purified
Host Species Rabbit
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

APPA6826

 
pierce biotechnology inc
PA116826
Unlabeled 50μL Each for $448.05
Description & Specifications

Specifications

Antigen CENPF
Cross Reactivity Bovine
Cross Reactivity Human
Format Purified
Host Species Rabbit
Immunogen A bacterial fusion protein from the C-terminus of CENP-F.
Isotype IgG
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Storage Requirements Avoid Freeze/Thaw Cycles

CENPF Polyclonal Ab PA1-16826 from Thermo Scientific™ specifically detects CENPF in Bv and Hu samples, and it is validated for Flow Cytometry, ICC, IF, IP and WB

The kinetophore is a macromolecular structure that is associated with the centromeres of chromosomes and is responsible for establishing and maintaining the connections with the microtubules of the mitotic spindle. The proteins that reside within the chromosomal domain fall into two broad classes: those that associate with the centromere throughout the cell cycle and those that are found at the centromere in a cell cycle specific manner. Members of the first class include CENP-A, CENP-B, and CENP-C. The members of the second class include several ubiquitous cellular proteins such as calmodulin, tubulin, dynein and p34 cdc2 and also CENP-E and CENP-F. CENP-F appears to be the earliest member to interact with the centromere-kinetochore complex. CENP-F also shows a cell-cycle distribution that is both temporally regulated and diverse in terms of the structural components it localizes to and therefore may play a role in several mitotic events.
Host Species: Rabbit
Cross Reactivity: Bovine, Human
Isotype: IgG
Immunogen: A bacterial fusion protein from the C-terminus of CENP-F.
Formulation: 50μL purified Antibody (1.25mg/mL) in PBS, with 50% glycerol and 0.05% sodium azide;
Synonyms: AH Antigen, CENF, CENP F Kinetochore Protein, Centromere Protein F, Mitosin, CENPF
Uniprot Accession: P49454
Gene Symbol: CENPF
Entrez GeneID: 1063
PA1-16826 detects CENPF in Bovine and Human samples. PA1-16826 has been successfully used in Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation and Western Blot procedures. PA1-16826 The immunogen is A bacterial fusion protein from the C-terminus of CENP-F..

Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly